Learn More
Bon Opus Biosciences Recombinant Human Apolipoprotein E/ ApoE (C-His, Expi293) -500ug
SDP
Catalog No. 502080193
Encompass_Preferred
Recombinant Human Apolipoprotein E is produced by Human 293 Cells and the target gene encoding Lys19?His317 is expressed with a 6His tag at the C?terminus. Expression system: Human 293 Cells. Accession# P02649. Formulation: Lyophilized frum a 0.2 um filtered solution of 2umM PBS, 5mM CHAPS, 2mM DDT, pH7.4. Bon Opus Cat# BP120
Catalog No. 50-208-0193 Supplier Bon Opus Biosciences Supplier No. BP120500UG
Recombinant Human Apolipoprotein E is produced by Human 293 Cells and the target gene encoding Lys19?His317 is expressed with a 6His tag at the C?terminus. Expression system: Human 293 Cells. Accession# P02649. Formulation: Lyophilized frum a 0.2 um filtered solution of 2umM PBS, 5mM CHAPS, 2mM DDT, pH7.4. Bon Opus Cat# BP120