Learn More
Bon Opus Biosciences Recombinant Human Apolipoprotein D/ApoD (C-6His) 500ug
SDP
Supplier:  Bon Opus Biosciences BP207500UG
Encompass_Preferred
Recombinant Human Apolipoprotein D is produced by our Mammalian expression system and the target gene encoding Glu21-Ser189 is expressed with a 6His tag at the C-terminus. Accession# P05090. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Bon Opus Cat# C556
Catalog No. 50-217-2752