Learn More
Bon Opus Biosciences Recombinant Human Arginase-2/ARG2 (C-6His) 1mg
SDP
Supplier:  Bon Opus Biosciences CE711MG
Encompass_Preferred
Recombinant Human Arginase-2, mitochondrial is produced by our E.coli expression system and the target gene encoding His24-Gly330 is expressed with a 6His tag at the C-terminus. Accession# P78540. Formulation: Supplied as a 0.2 μm filtered solution of 10mM TrisHCl,10mM NaCl,1mM β-mercaptoethanol,pH7.5. Bon Opus Cat# CE71
Catalog No. 50-217-2781