Learn More
Bon Opus Biosciences Recombinant Human Tyrosine-protein Kinase Receptor UFO/AXL (C-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP078500UG
Encompass_Preferred
Recombinant Human Tyrosine-protein Kinase Receptor UFO is produced by Human 293 Cells. The target gene encoding A26-P449 is expressed with a 8His tag at the C terminus. Expression system: Human 293 Cells. Accession# P30530. Formulation: Supplied as a 0.2 um filtered solution of PBS, pH7.2. Bon Opus Cat# BP078
Catalog No. 50-208-0153