Learn More
Bon Opus Biosciences Recombinant Human Beta-2 microglobulin/B2M (C-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP162500UG
Encompass_Preferred
Recombinant Human Beta-2 microglobulin is produced by our Mammalian expression system and the target gene encoding Ile21-Met119 is expressed with a 6His tag at the C-terminus. Expression system: Human 293 Cells. Accession# P61769. Formulation: Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Bon Opus Cat# BP162
Catalog No. 50-208-0258