Learn More
Bon Opus Biosciences Recombinant Human B7-2/CD86 (C-His)
SDP
Supplier:  Bon Opus Biosciences BP01450UG
Encompass_Preferred
Recombinant Human CD86 is produced by our Mammalian expression system and the target gene encoding Ala24-Pro247 is expressed with a 6His tag at the C-terminus. Accession# P42081. Bon Opus Cat# BP014
Catalog No. 50-203-1801