Learn More
Bon Opus Biosciences Recombinant Human BCMA (N-6His-Flag) -50ug
SDP
Catalog No. 502080326
Encompass_Preferred
Recombinant Human Tumor Necrosis Factor Receptor Superfamily Member 17 is produced by our mammalian expression system and the target gene encoding Met1-Ala54 is expressed with a 6His, Flag tag at the N-terminus. Expression system: Human Cells. Accession# Q02223. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Bon Opus Cat# C01B
Catalog No. 50-208-0326 Supplier Bon Opus Biosciences Supplier No. C01B50UG
Recombinant Human Tumor Necrosis Factor Receptor Superfamily Member 17 is produced by our mammalian expression system and the target gene encoding Met1-Ala54 is expressed with a 6His, Flag tag at the N-terminus. Expression system: Human Cells. Accession# Q02223. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Bon Opus Cat# C01B