Learn More
Bon Opus Biosciences Recombinant Human Baculoviral IAP Repeat-Containing Protein 5/BIRC5/Survivin 500ug
SDP
Supplier:  Bon Opus Biosciences CE87500UG
Encompass_Preferred
Recombinant Human BIRC5 is produced by our E.coli expression system and the target gene encoding Met1-Asp142 is expressed. Accession# O15392. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM Tris, 0.1M NaCl, pH 7.5. Bon Opus Cat# CE87
Catalog No. 50-217-2782
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.