Learn More
Lonza Walkersville Hu BM Stromal Cells 5M
Catalog No. 5000496336
Encompass
When bone marrow cells are put into culture under specific conditions a stromal layer of adherent cells developes The stromal layer is composed of many cell types including fibroblasts mesenchymal stem cells adipocytes endothelial cells and macrophages These stromal cells function as a feeder layer for hematopetic progenitors and stem cells allowing proliferation and differentiation of progenitors and stem cells to continue for weeks in these cultures with no addition of exogenous cytokines This long term culture system allows researchers to study an in vitro model of the bone marrow microenvironment including cell-cell interactions adhesion molecules and cytokine secretion These and many other factors allow for the tight regulation of blood cell production progenitor cell commitment and differentiation stem cell renewal and LTC-IC assays Available as non-irradiated only HIV-1 hepatitis B and hepatitis C are not detected for all donors and/or cell lots
Catalog No. 50-004-96336 Supplier Lonza Walkersville Supplier No. 2M302
When bone marrow cells are put into culture under specific conditions a stromal layer of adherent cells developes The stromal layer is composed of many cell types including fibroblasts mesenchymal stem cells adipocytes endothelial cells and macrophages These stromal cells function as a feeder layer for hematopetic progenitors and stem cells allowing proliferation and differentiation of progenitors and stem cells to continue for weeks in these cultures with no addition of exogenous cytokines This long term culture system allows researchers to study an in vitro model of the bone marrow microenvironment including cell-cell interactions adhesion molecules and cytokine secretion These and many other factors allow for the tight regulation of blood cell production progenitor cell commitment and differentiation stem cell renewal and LTC-IC assays Available as non-irradiated only HIV-1 hepatitis B and hepatitis C are not detected for all donors and/or cell lots