Learn More
Bon Opus Biosciences Recombinant Human BTLA (C-mFc) -50ug
SDP
Supplier:  Bon Opus Biosciences CW4750UG
Encompass_Preferred
Recombinant Human B- and T-Lymphocyte Attenuator is produced by our Mammalian expression system and the target gene encoding Lys31-Leu150 is expressed with a mFc tag at the C-terminus. Expression system: Human Cells. Accession# Q7Z6A9-2. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Bon Opus Cat# CW47
Catalog No. 50-208-0430