Learn More
Bon Opus Biosciences Recombinant Human Butyrophilin Subfamily 2 Member A2/BTN2A2 (C-Fc) 1mg
SDP
Supplier:  Bon Opus Biosciences CU441MG
Encompass_Preferred
Recombinant Human Butyrophilin Subfamily 2 Member A2 is produced by our expression system and the target gene encoding Gln33-Val237 is expressed with a C-Fc tag. Accession# Q8WVV5. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH7.4. Bon Opus Cat# CU44
Catalog No. 50-217-2830