Learn More
Bon Opus Biosciences Recombinant Human C5a -500ug
SDP
Supplier:  Bon Opus Biosciences BP046500UG
Encompass_Preferred
Recombinant Human C5a is produced by E.coli. The target gene encoding T678-R751 is expressed. Expression system: E. coli. Accession# P01031. Formulation: Lyophilized frum a 0.2 um filtered solution of 2umM Tris-HCl, 30umM NaCl. pH8.0. Bon Opus Cat# BP046
Catalog No. 50-208-0123