Learn More
Bon Opus Biosciences Recombinant Human Calmodulin/CALM1
SDP
Supplier:  Bon Opus Biosciences C91150UG
Encompass_Preferred
Recombinant Human Calmodulin is produced by our E.coli expression system and the target gene encoding Met1-Lys149 is expressed. Bon Opus Cat. #C911 Accession# P62158. Bon Opus Cat# C911
Catalog No. 50-203-2573