Learn More
Bon Opus Biosciences Recombinant Human CaM Kinase I/CAMK1 (C-6His) 1mg
SDP
Supplier:  Bon Opus Biosciences C9371MG
Encompass_Preferred
Recombinant Human CaM Kinase I is produced by our Mammalian expression system and the target gene encoding Met1-Leu370 is expressed with a 6His tag at the C-terminus. Accession# Q14012. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB,150mM NaCl,pH7.4. Bon Opus Cat# C937
Catalog No. 50-217-2764