Learn More
Bon Opus Biosciences Recombinant Human Caspase-14/CASP14 (C-6His) 500ug
SDP
Supplier:  Bon Opus Biosciences CG19500UG
Encompass_Preferred
Recombinant Human Caspase-14 is produced by our E.coli expression system and the target gene encoding Ser2-Gln242 is expressed with a 6His tag at the C-terminus. Accession# P31944. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB, 150mM NaCl,pH7.4. Bon Opus Cat# CG19
Catalog No. 50-217-2786