Learn More
Lonza Walkersville Hu CB CD34 Stm/Prog Cells 1M
Supplier:  Lonza Walkersville 2C101
Encompass
Cord blood CD34 cells are known to contain hematopoietic stem and progenitor cells and are ideal for studies relating to transplantation and stem cells as cord blood derived cells tend to be more na ve than similar cells derived from other tissue sources High Purity and ViabilityLonza cord blood CD34 cells are isolated from cord blood mononuclear cells (MNCs) via positive immunomagnetic separation and are guaranteed to have a purity of 90% as indicated by CD34 expressing cells They are guaranteed to contain 1 million viable cells per ampoule HIV-1 and hepatitis B are not detected for all donors and/or cell lots A Certificate of Analysis is provided for each cell lot purchased Choices in Tissue Source - Cord blood CD34 cells tend to be more naive than hematopoietic stem cells from other tissue sources Thus they can be a great tool when trying to bypass HLA mismatch issues
Catalog No. 50-004-96416