Learn More
Bon Opus Biosciences Recombinant Human Coiled-Coil Domain-Containing Protein 134/CCDC134 (C-6His) 1mg
SDP
Supplier:  Bon Opus Biosciences CI221MG
Encompass_Preferred
Recombinant Human CCDC134 is produced by our Mammalian expression system and the target gene encoding Thr23-Leu229 is expressed with a 6His tag at the C-terminus. Accession# Q9H6E4. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB,150mM NaCl, pH7.4. Bon Opus Cat# CI22
Catalog No. 50-217-2800