Learn More
Bon Opus Biosciences Recombinant Human CCL20 (C-hFc) - Source: Human Cells
SDP
Catalog No. NC2733787
Encompass_Preferred
Recombinant Human CCL20 (C-hFc) is produced by Human Cells. The target gene encoding Ala27-Met96 is expressed with a C-hFc tag. Accession#AAH20698.1. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4.
Catalog No. NC2733787 Supplier Bon Opus Biosciences Supplier No. BP30950UG
Recombinant Human CCL20 (C-hFc) is produced by Human Cells. The target gene encoding Ala27-Met96 is expressed with a C-hFc tag. Accession#AAH20698.1. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4.