Learn More
Bon Opus Biosciences Recombinant Human IL-3R alpha/CD123 (C-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP064500UG
Encompass_Preferred
Recombinant Human IL-3R alpha is produced by Human 293 Cells. The target gene encoding K20-R305 is expressed with a 10His tag at the C terminus. Expression system: Human 293 Cells. Accession# P26951. Formulation: Lyophilized frum a 0.2 um filtered solution of PBS, pH 7.4. Bon Opus Cat# BP064
Catalog No. 50-208-0139