Learn More
Bon Opus Biosciences Recombinant Human Coagulation Factor III/Tissue Factor/CD142 (C-6His) 500ug
SDP
Supplier:  Bon Opus Biosciences C479500UG
Encompass_Preferred
Recombinant Human Coagulation Factor III is produced by our Mammalian expression system and the target gene encoding Gly34-Glu251 is expressed with a 6His tag at the C-terminus. Accession# P13726. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Bon Opus Cat# C479
Catalog No. 50-217-2748