Learn More
Bon Opus Biosciences Recombinant Human CD160 (C-Fc) -50ug
SDP
Supplier:  Bon Opus Biosciences CW0450UG
Encompass_Preferred
Recombinant Human CD160 is produced by our Mammalian expression system and the target gene encoding Ile27-Ser159 is expressed with a Fc tag at the C-terminus. Expression system: Human Cells. Accession# O95971. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Bon Opus Cat# CW04
Catalog No. 50-208-0407