Learn More
Lonza Walkersville Hu CD19 B-Cells Neg 10M
Catalog No. 5000496223
Encompass
Leukapheresed CD19 B cells are negatively selected from leukapheresed peripheral blood MNCs using an immunomagnetic labeling system to remove unwanted cell types leaving the B cells untouched Cells are cryopreserved in Lonza Cryo Media in a cryovial Cell purity is determined post thaw using a FACS assay Count and viability is determined using AO/PI Cells are collected from healthy donors following IRB protocols
Catalog No. 50-004-96223 Supplier Lonza Walkersville Supplier No. 4W601
Leukapheresed CD19 B cells are negatively selected from leukapheresed peripheral blood MNCs using an immunomagnetic labeling system to remove unwanted cell types leaving the B cells untouched Cells are cryopreserved in Lonza Cryo Media in a cryovial Cell purity is determined post thaw using a FACS assay Count and viability is determined using AO/PI Cells are collected from healthy donors following IRB protocols