Learn More
Lonza Walkersville Hu CD19 B-Cells Neg 10M
Catalog No. 5000496302
Encompass
CD19 B cells are negatively selected from leukapheresed peripheral blood MNCs using an immunomagnetic labeling system to remove unwanted cell types Cells are cryopreserved in Lonza Cryo Media in a cryovial Cell purity is determined post thaw using a FACS assay Count and viability is determined using hemacytometer (trypan blue method) Cells are collected from healthy donors following IRB protocols
Catalog No. 50-004-96302 Supplier Lonza Walkersville Supplier No. 2W601
CD19 B cells are negatively selected from leukapheresed peripheral blood MNCs using an immunomagnetic labeling system to remove unwanted cell types Cells are cryopreserved in Lonza Cryo Media in a cryovial Cell purity is determined post thaw using a FACS assay Count and viability is determined using hemacytometer (trypan blue method) Cells are collected from healthy donors following IRB protocols