Learn More
Bon Opus Biosciences Recombinant Human CD200 (C-His)
SDP
Supplier:  Bon Opus Biosciences BP00450UG
Encompass_Preferred
Recombinant Human CD200 is produced by human 293 cells. The target gene encoding Gln31-Gly232 is expressed with a 6His tag at the C-terminus. Accession# P41217. Bon Opus Cat# BP004
Catalog No. 50-203-1792