Learn More
Bon Opus Biosciences Recombinant Human CD200 (C-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP004500UG
Encompass_Preferred
Recombinant Human CD200 is produced by human 293 cells. The target gene encoding Gln31-Gly232 is expressed with a 6His tag at the C-terminus. Expression system: Human 293 Cells. Accession# P41217. Formulation: Lyophilized frum a 0.2 um filtered solution of 2umM PB, 15umM NaCl, pH 7.4. Bon Opus Cat# BP004
Catalog No. 50-208-0085