Learn More
Bon Opus Biosciences Recombinant Human Siglec-2/CD22 (C-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP063500UG
Encompass_Preferred
Recombinant Human Siglec-2 is produced by Human 293 Cells. The target gene encoding D20-R687 is expressed with a 10His tag at the C terminus. Expression system: Human 293 Cells. Accession# P20273. Formulation: Lyophilized frum a 0.2 um filtered solution of PBS, pH 7.4. Bon Opus Cat# BP063
Catalog No. 50-208-0138