Learn More
Bon Opus Biosciences Recombinant Human IL-2RA/CD25 (C-hFc) -500ug
SDP
Supplier:  Bon Opus Biosciences BP149500UG
Encompass_Preferred
Recombinant Human Interleukin-2 Receptor subunit alpha is produced by our Mammalian expression system and the target gene encoding Glu22-Cys213 is expressed with a Fc tag at the C-terminus. Expression system: Human 293 Cells. Accession# P01589. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# BP149
Catalog No. 50-208-0236