Learn More
Bon Opus Biosciences Recombinant Human BTN3A1/CD277 (C-hFc) -500ug
SDP
Supplier:  Bon Opus Biosciences BP185500UG
Encompass_Preferred
Recombinant Human Butyrophilin Subfamily 3 Member A1 is produced by our Mammalian expression system and the target gene encoding Gln30-Gly254 is expressed with a Fc tag at the C-terminus. Expression system: Human 293 Cells. Accession# O00481. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, 5% Trehalose, 0.01%Tween 80, pH7.4. Bon Opus Cat# BP185
Catalog No. 50-208-0296