Learn More
Bon Opus Biosciences Recombinant Human CD27L (C-mFc) -50ug
SDP
Supplier:  Bon Opus Biosciences CW4850UG
Encompass_Preferred
Recombinant Human CD27 Ligand/CD70 is produced by our Mammalian expression system and the target gene encoding Gln45-Pro193 is expressed with a mFc tag at the C-terminus. Expression system: Human Cells. Accession# P32970. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Bon Opus Cat# CW48
Catalog No. 50-208-0431