Learn More
Apexbio Technology LLC HU CD3 T CELL ISO NEG SEL-0.1g
SDP
Catalog No. 5000897347
Encompass_Preferred
Human CD3 T Cell Isolation Kit (Negative Selection) is designed for research in immunology and cell biology focusing on T cell studies Originating from human peripheral blood its mechanism involves negative selection to isolate untouched CD3 T cells by depleting non-target cells using antibodies and magnetic particles These cells are typically used in vitro for functional studies including proliferation and cytokine production assays and in vivo for animal model research The kit is valuable for drug screening and immunotherapy research where the purity of T cells is crucial Experimental concentrations typically depend on the specific research design
Catalog No. 50-008-97347 Supplier Apexbio Technology LLC Supplier No. CS1001100
Human CD3 T Cell Isolation Kit (Negative Selection) is designed for research in immunology and cell biology focusing on T cell studies Originating from human peripheral blood its mechanism involves negative selection to isolate untouched CD3 T cells by depleting non-target cells using antibodies and magnetic particles These cells are typically used in vitro for functional studies including proliferation and cytokine production assays and in vivo for animal model research The kit is valuable for drug screening and immunotherapy research where the purity of T cells is crucial Experimental concentrations typically depend on the specific research design