Learn More
Abcam Human CD30-L ELISA Kit (TNFSF8), 1 x 96 Tests
Supplier:  Abcam AB2523621X96TESTS
Encompass_Preferred
Human CD30-L ELISA Kit (TNFSF8) is a single-wash 90-min Simplestep used to quantify Human CD30-L (TNFSF8) with a sensitivity of 0.06 ng/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2748830