Learn More
Bon Opus Biosciences Recombinant Human Siglec‑3/CD33 (C-hFc/His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP175500UG
Encompass_Preferred
Recombinant Human Sialic Acid Binding Ig-like Lectin 3 is produced by our Mammalian expression system and the target gene encoding Asp18-His259 is expressed with a Fc / 6His tag at the C-terminus. Expression system: Human 293 Cells. Accession# P20138. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, 5% Trehalose, 0.01%Tween 80, pH7.2. Bon Opus Cat# BP175
Catalog No. 50-208-0280