Learn More
Lonza Walkersville Hu CD34 Prog Cells from Mobil
Catalog No. 5000496344
Encompass
Mobilized peripheral blood CD34 hematopoietic stem and progenitor cells are positively selected using a immunomagnetic CD34 microbead labeling system Cells are formulated in Lonza Fresh Media and shipped in a cryovial Cell purity is determined using a FACS assay Count and viability is determined using AO/PI Cells are collected from healthy donors following IRB protocols
Catalog No. 50-004-96344 Supplier Lonza Walkersville Supplier No. 3Y101C
Mobilized peripheral blood CD34 hematopoietic stem and progenitor cells are positively selected using a immunomagnetic CD34 microbead labeling system Cells are formulated in Lonza Fresh Media and shipped in a cryovial Cell purity is determined using a FACS assay Count and viability is determined using AO/PI Cells are collected from healthy donors following IRB protocols