Learn More
Lonza Walkersville Hu CD34 Prog Cells Pos 10M
Catalog No. 5000496291
Encompass
Mobilized peripheral blood CD34 hematopoietic stem and progenitor cells are positively selected using a immunomagnetic CD34 microbead labeling system Cells are cryopreserved in Lonza Cryo Media in a cryovial Cell purity is determined post thaw using a FACS assay Count and viability is determined using AO/PI Cells are collected from healthy donors following IRB protocols Manufactured by AllCells
Catalog No. 50-004-96291 Supplier Lonza Walkersville Supplier No. 4Y101E
Add to Cart
Add to Cart
Mobilized peripheral blood CD34 hematopoietic stem and progenitor cells are positively selected using a immunomagnetic CD34 microbead labeling system Cells are cryopreserved in Lonza Cryo Media in a cryovial Cell purity is determined post thaw using a FACS assay Count and viability is determined using AO/PI Cells are collected from healthy donors following IRB protocols Manufactured by AllCells