Learn More
Lonza Walkersville Hu CD34 Prog Cells Pos 5M
Catalog No. 5000496345
Encompass
Mobilized peripheral blood CD34 hematopoietic stem and progenitor cells are positively selected using a immunomagnetic CD34 microbead labeling system Cells are cryopreserved in Lonza Cryo Media in a cryovial Cell purity is determined post thaw using a FACS assay Count and viability is determined using hemacytometer (trypan blue method) Cells are collected from healthy donors following IRB protocols
Catalog No. 50-004-96345 Supplier Lonza Walkersville Supplier No. 2Y101D
Mobilized peripheral blood CD34 hematopoietic stem and progenitor cells are positively selected using a immunomagnetic CD34 microbead labeling system Cells are cryopreserved in Lonza Cryo Media in a cryovial Cell purity is determined post thaw using a FACS assay Count and viability is determined using hemacytometer (trypan blue method) Cells are collected from healthy donors following IRB protocols