Learn More
Bon Opus Biosciences Recombinant Human CD3D/CD3E Heterodimer (C-6His) -50ug
SDP
Supplier:  Bon Opus Biosciences C01S50UG
Encompass_Preferred
Recombinant Human CD3D/CD3E is produced by our Mammalian expression system and the target geneencoding Phe22-Ala105/Asp23-Asp126 is expressed with a 6His tag at the C-terminus. Expression system: Human Cells. Accession# P04234/P07766. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Bon Opus Cat# C01S
Catalog No. 50-208-0336