Learn More
Bon Opus Biosciences Recombinant Human CD3E (C-hFc) -500ug
SDP
Supplier:  Bon Opus Biosciences BP187500UG
Encompass_Preferred
Recombinant Human T-cell surface glycoprotein CD3 epsilon chain is produced by our Mammalian expression system and the target gene encoding Asp23-Asp126 is expressed with a Fc tag at the C-terminus. Expression system: Human 293 Cells. Accession# P07766. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, 0.01%Tween 80, pH 7.4. Bon Opus Cat# BP187
Catalog No. 50-208-0300