Learn More
Apexbio Technology LLC HU CD4 T CELL ISO NEG SEL-0.1g
SDP
Catalog No. 5000897326
Encompass_Preferred
Human CD4 T Cell Isolation Kit (Negative Selection) is a key tool in immunology research designed to isolate untouched CD4 T cells from human peripheral blood mononuclear cells Originating from the principle of negative selection the kit removes unwanted cells using antibodies targeting non-CD4 cells allowing the untouched CD4 cells to remain This method maintains cell integrity and functionality The kit is commonly used in vitro with activity viable in nanomolar to low micromolar ranges depending on the assay conditions It is widely utilized in applications such as studying T cell behavior in various diseases drug screening and evaluating immune responses in cell models and pre-clinical animal studies Experimental concentrations typically depend on the specific research design ensuring flexibility for diverse research needs
Catalog No. 50-008-97326 Supplier Apexbio Technology LLC Supplier No. CS1002100
Human CD4 T Cell Isolation Kit (Negative Selection) is a key tool in immunology research designed to isolate untouched CD4 T cells from human peripheral blood mononuclear cells Originating from the principle of negative selection the kit removes unwanted cells using antibodies targeting non-CD4 cells allowing the untouched CD4 cells to remain This method maintains cell integrity and functionality The kit is commonly used in vitro with activity viable in nanomolar to low micromolar ranges depending on the assay conditions It is widely utilized in applications such as studying T cell behavior in various diseases drug screening and evaluating immune responses in cell models and pre-clinical animal studies Experimental concentrations typically depend on the specific research design ensuring flexibility for diverse research needs