Learn More
Bon Opus Biosciences Recombinant Human CD46/MCP (C-6His) 500ug
SDP
Supplier:  Bon Opus Biosciences C320500UG
Encompass_Preferred
Recombinant Human CD46 is produced by our Mammalian expression system and the target gene encoding Cys35-Asp328 is expressed with a 6His tag at the C-terminus. Accession# P15529. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Bon Opus Cat# C320
Catalog No. 50-217-2734