Learn More
Bon Opus Biosciences Recombinant Human CD47 (C-His) -500ug
SDP
Catalog No. 502080226
Encompass_Preferred
Recombinant Human CD47 is produced by Human 293 Cells. The target gene encoding Gln19-Pro139 is expressed with a 6His tag at the C terminus. Expression system: Human 293 Cells. Accession# Q08722. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Bon Opus Cat# BP142
Catalog No. 50-208-0226 Supplier Bon Opus Biosciences Supplier No. BP142500UG
Add to Cart
Add to Cart
Recombinant Human CD47 is produced by Human 293 Cells. The target gene encoding Gln19-Pro139 is expressed with a 6His tag at the C terminus. Expression system: Human 293 Cells. Accession# Q08722. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Bon Opus Cat# BP142