Learn More
Bon Opus Biosciences Recombinant Human CD47 (C-mFc) -50ug
SDP
Supplier:  Bon Opus Biosciences CW2850UG
Encompass_Preferred
Recombinant Human Leukocyte Surface Antigen CD47 is produced by our Mammalian expression systemand the target gene encoding Gln19-Pro139 is expressed with a mFc tag at the C-terminus. Expression system: Human Cells. Accession# Q08722. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Bon Opus Cat# CW28
Catalog No. 50-208-0416