Learn More
Bon Opus Biosciences Recombinant Human CD47/IAP/OA3 (C-6His) 1mg
SDP
Catalog No. 502172735
Encompass_Preferred
Recombinant Human CD47 is produced by our Mammalian expression system and the target gene encoding Gln19-Pro139 is expressed with a 6His tag at the C-terminus. Accession# Q08722. Formulation: Lyophilized from a 0.2 μm filtered solution of 10mM Tris-Citrate, 150mM NaCl, pH 8.0. Bon Opus Cat# C321
Catalog No. 50-217-2735 Supplier Bon Opus Biosciences Supplier No. C3211MG
Recombinant Human CD47 is produced by our Mammalian expression system and the target gene encoding Gln19-Pro139 is expressed with a 6His tag at the C-terminus. Accession# Q08722. Formulation: Lyophilized from a 0.2 μm filtered solution of 10mM Tris-Citrate, 150mM NaCl, pH 8.0. Bon Opus Cat# C321