Learn More
Bon Opus Biosciences Biotinylated Recombinant Human E-selectin/ELAM1/CD62E (C-His) -250ug
SDP
Supplier:  Bon Opus Biosciences BP098250UG
Encompass_Preferred
Biotinylated Recombinant Human E-selectin is produced by Human 293 Cells. The target gene encoding W22-P556 is expressed with a 8His tag at the C terminus. Expression system: Human 293 Cells. Accession# P16582. Formulation: Lyophilized frum a 0.2 um filtered solution of PBS, pH7.2. Bon Opus Cat# BP098
Catalog No. 50-208-0173