Learn More
Bon Opus Biosciences Recombinant Human CD79A(C-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP158500UG
Encompass_Preferred
Recombinant Human CD79A is produced by our Mammalian expression system and the target gene encoding Leu33-Arg143 is expressed with a 6His tag at the C-terminus. Expression system: Human 293 Cells. Accession# P11912. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Bon Opus Cat# BP158
Catalog No. 50-208-0250