Learn More
Bon Opus Biosciences Recombinant Human CD79B (C-Fc) -50ug
SDP
Supplier:  Bon Opus Biosciences CW9650UG
Encompass_Preferred
Recombinant Human CD79B is produced by our Mammalian expression system and the target gene encoding Ala29-Asp159 is expressed with a Fc tag at the C-terminus. Expression system: Human Cells. Accession# P40259. Formulation: Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Bon Opus Cat# CW96
Catalog No. 50-208-0473