Learn More
Bon Opus Biosciences Recombinant Human B7-1/CD80 (C-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP015500UG
Encompass_Preferred
Recombinant Human CD80 is produced with our Mammalian expression system. The target protein is expressed with sequence (Val35-Asn242) of Human CD80 fused with a 6-His tag at the C-terminus. Expression system: Human 293 Cells. Accession# P33681. Formulation: Lyophilized frum a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# BP015
Catalog No. 50-208-0095