Learn More
Bon Opus Biosciences Recombinant Human Cadherin-1/E-cadherin/CDH1 (C-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP099500UG
Encompass_Preferred
Recombinant Human Cadherin-1 is produced by Human 293 Cells. The target gene encoding Q23-A709 is expressed with a 8His tag at the C terminus. Expression system: Human 293 Cells. Accession# P12830. Formulation: Lyophilized frum a 0.2 um filtered solution of PBS, pH7.2. Bon Opus Cat# BP099
Catalog No. 50-208-0174