Learn More
Bon Opus Biosciences Recombinant Human Cadherin-11/CDH11 (C-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP170500UG
Encompass_Preferred
Recombinant Human Cadherin-11 is produced by our Mammalian expression system and the target gene encoding Phe23-Thr617 is expressed with a 6His tag at the C-terminus. Expression system: Human 293 Cells. Accession# Q96CZ9. Formulation: Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Bon Opus Cat# BP170
Catalog No. 50-208-0270