Learn More
Bon Opus Biosciences Recombinant Human Cadherin-3/CDH3 (C-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP126500UG
Encompass_Preferred
Recombinant Human Cadherin-3 is produced by our Mammalian expression system and the target gene encoding Glu25-Gly654 is expressed with a 6His tag at the C-terminus. Expression system: Human 293 Cells. Accession# P22223. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# BP126
Catalog No. 50-208-0201