Learn More
Bon Opus Biosciences Recombinant Human Complement Factor H-related Protein 1/CFHR1(C-His) -50ug
SDP
Supplier:  Bon Opus Biosciences BP16450UG
Encompass_Preferred
Recombinant Human Complement Factor H-Related 1 is produced by our Mammalian expression system and the target gene encoding Glu19-Arg330 is expressed with a 6His tag at the C-terminus. Expression system: Human 293 Cells. Accession# Q03591. Formulation: Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, 0.01%Tween 80, pH 7.4. Bon Opus Cat# BP164
Catalog No. 50-208-0261